Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RSH2
- Entrez Gene ID
-
820619
- Gene Name
- RELA/SPOT homolog 2
- Gene Aliases
-
AT-RSH2, AT3G14050, ATRSH2, RELA-SPOT HOMOLOG 2, RELA/SPOT HOMOLOG 2, RELA/SPOT homolog 2
- Chromosome Location
-
Chr.3: 4650714 - 4654046 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_112259.5 |
NP_188021.1 |
5 - 6 |
2366 |
78 |
View Full Product Details