Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBP25
Entrez Gene ID
820662
Gene Name
ubiquitin-specific protease 25
Gene Aliases
AT3G14400, UBIQUITIN-SPECIFIC PROTEASE 25, ubiquitin-specific protease 25
Chromosome Location
Chr.3: 4811486 - 4815515 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112299.4 NP_566486.1 4 - 5 1116 119
GenBank mRNA AF302673.1 - 4 - 5 811 119
BT021128.1 - 4 - 5 832 119

View Full Product Details