Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP72A9
Entrez Gene ID
820691
Gene Name
cytochrome P450, family 72, subfamily A, polypeptide 9
Gene Aliases
''cytochrome P450, AT3G14630, cytochrome P450, family 72, polypeptide 9, polypeptide 9'', subfamily A
Chromosome Location
Chr.3: 4917327 - 4919702 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338127.1 NP_001319551.1 1 - 2 265 105
NM_112324.2 NP_188081.2 1 - 2 436 105

View Full Product Details