Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SHM3
Entrez Gene ID
829387
Gene Name
serine hydroxymethyltransferase 3
Gene Aliases
AT4G32520, AtSHMT3, L23H3.3, SERINE HYDROXYMETHYLTRANSFERASE 3, serine hydroxymethyltransferase 3
Chromosome Location
Chr.4: 15689436 - 15692989 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125626.1 NP_001119098.1 11 - 12 1390 140
NM_119404.3 NP_567895.1 11 - 12 1603 140

View Full Product Details