Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LUG
Entrez Gene ID
829390
Gene Name
transcriptional corepressor LEUNIG
Gene Aliases
AT4G32551, L23H3.30, L23H3_30, LEUNIG, RON2, ROTUNDA 2
Chromosome Location
Chr.4: 15707425 - 15713662 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203962.1 NP_001190891.1 7 - 8 1515 72
NM_119407.3 NP_567896.1 8 - 9 1457 72

View Full Product Details