Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
XI-I
Entrez Gene ID
829456
Gene Name
myosin
Gene Aliases
AT4G33200, ATXI-I, F4I10.130, F4I10_130, MYOSIN XI I, MYOSIN XI-15, XI-15
Chromosome Location
Chr.4: 16002466 - 16014981 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203968.1 NP_001190897.1 3 - 4 585 130
NM_001203969.1 NP_001190898.1 3 - 4 585 130
NM_001342203.1 NP_001328260.1 3 - 4 607 130
NM_119474.5 NP_195046.3 3 - 4 619 130
GenBank mRNA AK228969.1 - 3 - 4 606 130

View Full Product Details