Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KUP5
Entrez Gene ID
829492
Gene Name
K+ uptake permease 5
Gene Aliases
AT4G33530, K+ uptake permease 5, T16L1.20, T16L1_20
Chromosome Location
Chr.4: 16126179 - 16130608 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342219.1 NP_001329920.1 4 - 5 1402 139
NM_119508.4 NP_195079.2 5 - 6 1346 139
GenBank mRNA AY099556.1 - 5 - 6 1176 139
BT010357.1 - 5 - 6 1091 139

View Full Product Details