Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FUC95A
Entrez Gene ID
829575
Gene Name
1,2-alpha-L-fucosidase
Gene Aliases
ALTERED XYLOGLUCAN 8, AT4G34260, AXY8, F10M10.30, F10M10_30
Chromosome Location
Chr.4: 16397995 - 16402118 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119590.4 NP_195152.2 2 - 3 551 125
GenBank mRNA AY125494.1 - 2 - 3 420 125
BT002722.1 - 2 - 3 416 125

View Full Product Details