Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLP2
Entrez Gene ID
829650
Gene Name
subtilisin-like serine protease 2
Gene Aliases
AT4G34980, M4E13.40, M4E13_40, subtilisin-like serine protease 2
Chromosome Location
Chr.4: 16656622 - 16659486 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119664.3 NP_567972.1 1 - 1 2581 71
GenBank mRNA AY074375.1 - 1 - 1 2434 71

View Full Product Details