Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SDG37
- Entrez Gene ID
-
816300
- Gene Name
- SET domain group 37
- Gene Aliases
-
ASH1-related 1, ASHR1, AT2G17900, SET DOMAIN GROUP 37, SET domain group 37, T13L16.8, T13L16_8
- Chromosome Location
-
Chr.2: 7773220 - 7776825 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_179638.2 |
NP_849969.2 |
2 - 3 |
396 |
75 |
View Full Product Details