Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHT4;1
Entrez Gene ID
817515
Gene Name
phosphate transporter 4;1
Gene Aliases
ANTR1, AT2G29650, T27A16.25, T27A16_25, anion transporter 1, phosphate transporter 4;1
Chromosome Location
Chr.2: 12673099 - 12676092 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336220.1 NP_001325344.1 2 - 3 744 85
NM_128519.6 NP_180526.1 3 - 4 697 85
NM_179805.2 NP_850136.1 3 - 4 712 85
NM_201832.1 NP_973561.1 3 - 4 510 85
GenBank mRNA BT001983.1 - 3 - 4 671 85
BT008489.1 - 3 - 4 623 85

View Full Product Details