Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ATX1
- Entrez Gene ID
-
817721
- Gene Name
- homologue of trithorax
- Gene Aliases
-
AT2G31650, SDG27, SET DOMAIN PROTEIN 27, T9H9.17, T9H9_17, homologue of trithorax
- Chromosome Location
-
Chr.2: 13455272 - 13462431 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_179839.3 |
NP_850170.1 |
9 - 10 |
1804 |
65 |
View Full Product Details