Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAP
Entrez Gene ID
817747
Gene Name
RAP
Gene Aliases
AT2G31890, ATRAP, F20M17.7, F20M17_7
Chromosome Location
Chr.2: 13556500 - 13559880 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_179845.4 NP_850176.1 2 - 3 399 66
GenBank mRNA AY065010.1 - 2 - 3 286 66
AY129492.1 - 2 - 3 234 66

View Full Product Details