Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UVR3
Entrez Gene ID
820804
Gene Name
DNA photolyase family protein
Gene Aliases
6-4 PHOTOLYASE, AT3G15620, UV REPAIR DEFECTIVE 3
Chromosome Location
Chr.3: 5293256 - 5296811 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112432.2 NP_566520.1 11 - 12 1573 131

View Full Product Details