Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LPD1
Entrez Gene ID
820951
Gene Name
lipoamide dehydrogenase 1
Gene Aliases
AT3G16950, LIPOAMIDE DEHYDROGENASE, lipoamide dehydrogenase 1, ptlpd1
Chromosome Location
Chr.3: 5786298 - 5790536 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084696.1 NP_001078165.1 14 - 15 1797 61
NM_112571.3 NP_566562.1 14 - 15 1808 61
GenBank mRNA AF228637.1 - 14 - 15 1761 61
AY050376.1 - 14 - 15 1697 61
AY120695.1 - 14 - 15 1655 61

View Full Product Details