Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scpl33
Entrez Gene ID
820975
Gene Name
serine carboxypeptidase-like 33
Gene Aliases
AT3G17180, serine carboxypeptidase-like 33
Chromosome Location
Chr.3: 5855632 - 5859189 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338277.1 NP_001326250.1 3 - 4 480 104
NM_112594.2 NP_188343.1 3 - 4 512 104

View Full Product Details