Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- AT3G17670
- Entrez Gene ID
-
821034
- Gene Name
- tetratricopeptide repeat (TPR)-containing protein
- Gene Aliases
-
AT3G17670, ENA, ENHANCER OF ATNSI ACTIVITY
- Chromosome Location
-
Chr.3: 6040797 - 6042196 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_112648.3 |
NP_188394.2 |
3 - 4 |
604 |
153 |
View Full Product Details