Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MPK9
Entrez Gene ID
821329
Gene Name
MAP kinase 9
Gene Aliases
AT3G18040, MAP kinase 9
Chromosome Location
Chr.3: 6174239 - 6178534 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338319.1 NP_001327296.1 9 - 10 1798 97
NM_001338320.1 NP_001327298.1 9 - 10 1798 97
NM_001338321.1 NP_001327297.1 9 - 10 1718 97
NM_112686.4 NP_566595.1 9 - 10 1318 97
NM_202602.2 NP_974331.1 8 - 9 1841 97

View Full Product Details