Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NRPD2B
Entrez Gene ID
821334
Gene Name
nuclear RNA polymerase D2B
Gene Aliases
AT3G18090, nuclear RNA polymerase D2B
Chromosome Location
Chr.3: 6194509 - 6200424 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338327.1 NP_001326261.1 3 - 4 855 76
NM_001338328.1 NP_001326262.1 3 - 4 848 76
NM_112691.4 NP_188437.2 3 - 4 848 76

View Full Product Details