Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATHXK4
Entrez Gene ID
821543
Gene Name
Hexokinase
Gene Aliases
AT3G20040, HEXOKINASE-LIKE 2, HKL2
Chromosome Location
Chr.3: 6994770 - 6998185 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112895.3 NP_188639.2 5 - 6 1382 63

View Full Product Details