Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCI15
Entrez Gene ID
821576
Gene Name
trigalactosyldiacylglycerol2
Gene Aliases
AT3G20320, ATP-binding cassette I15, TGD2, trigalactosyldiacylglycerol2
Chromosome Location
Chr.3: 7087445 - 7089927 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338475.1 NP_001319601.1 2 - 3 693 134
NM_112923.4 NP_566659.1 2 - 3 693 134
NM_202616.1 NP_974345.1 1 - 2 703 134
GenBank mRNA AY048230.1 - 2 - 3 628 134
AY094020.1 - 1 - 2 511 134

View Full Product Details