Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PDIL5-3
- Entrez Gene ID
-
821603
- Gene Name
- PDI-like 5-3
- Gene Aliases
-
ARABIDOPSIS THALIANA PROTEIN DISULFIDE ISOMERASE 12, AT3G20560, ATPDI12, ATPDIL5-3, PDI-like 5-3, PDI12, PROTEIN DISULFIDE ISOMERASE 12
- Chromosome Location
-
Chr.3: 7182013 - 7186567 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_112948.3 |
NP_566664.1 |
7 - 8 |
741 |
91 |
GenBank mRNA |
AY050453.1 |
- |
7 - 8 |
719 |
91 |
View Full Product Details