Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PLP4
- Entrez Gene ID
-
829859
- Gene Name
- PATATIN-like protein 4
- Gene Aliases
-
AP22.16, AP22_16, AT4G37050, AtPLAIVC, PATATIN-like protein 4, PLA V, PLAIII{beta}, patatin-related phospholipase III beta, phospholipase A IVC
- Chromosome Location
-
Chr.4: 17457135 - 17459770 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_119868.3 |
NP_195422.3 |
5 - 6 |
1172 |
119 |
View Full Product Details