Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scpl46
Entrez Gene ID
817918
Gene Name
serine carboxypeptidase-like 46
Gene Aliases
AT2G33530, F4P9.30, F4P9_30, serine carboxypeptidase-like 46
Chromosome Location
Chr.2: 14197646 - 14200674 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_179881.4 NP_850212.1 9 - 10 1390 73
GenBank mRNA AY074317.1 - 9 - 10 1273 73

View Full Product Details