Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAX1
Entrez Gene ID
818395
Gene Name
cation exchanger 1
Gene Aliases
AT2G38170, ATCAX1, F16M14.10, F16M14_10, HIGH AFFINITY CALCIUM ANTIPORTER CAX1, RARE COLD INDUCIBLE 4, RCI4, cation exchanger 1
Chromosome Location
Chr.2: 15989083 - 15993326 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129373.4 NP_181352.1 9 - 10 1464 111
NM_179961.2 NP_850292.2 8 - 9 1518 111
GenBank mRNA AF372938.1 - 9 - 10 1411 111

View Full Product Details