Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- FAC1
- Entrez Gene ID
-
818408
- Gene Name
- AMP deaminase, putative / myoadenylate deaminase
- Gene Aliases
-
ADENOSINE 5'-MONOPHOSPHATE DEAMINASE, AT2G38280, ATAMPD, EMBRYONIC FACTOR1, F16M14.21, F16M14_21
- Chromosome Location
-
Chr.2: 16033292 - 16038972 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_179963.3 |
NP_850294.1 |
19 - 20 |
2743 |
87 |
View Full Product Details