Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAC1
Entrez Gene ID
818408
Gene Name
AMP deaminase, putative / myoadenylate deaminase
Gene Aliases
ADENOSINE 5'-MONOPHOSPHATE DEAMINASE, AT2G38280, ATAMPD, EMBRYONIC FACTOR1, F16M14.21, F16M14_21
Chromosome Location
Chr.2: 16033292 - 16038972 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129384.3 NP_565886.1 6 - 7 1246 133
NM_179963.3 NP_850294.1 6 - 7 1246 133
GenBank mRNA AY056301.1 - 6 - 7 1150 133
AY133852.1 - 6 - 7 1067 133

View Full Product Details