Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RCA
Entrez Gene ID
818558
Gene Name
rubisco activase
Gene Aliases
AT2G39730, RUBISCO ACTIVASE, T5I7.18, rubisco activase
Chromosome Location
Chr.2: 16570746 - 16573692 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129531.3 NP_565913.1 2 - 3 707 82
NM_179989.3 NP_850320.1 2 - 3 707 82
NM_179990.1 NP_850321.1 2 - 3 534 82
GenBank mRNA AF325049.2 - 2 - 3 448 82
AY052703.1 - 2 - 3 437 82
AY056108.1 - 2 - 3 448 82
AY088487.1 - 2 - 3 563 82
BT000710.1 - 2 - 3 445 82

View Full Product Details