Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RCA
Entrez Gene ID
818558
Gene Name
rubisco activase
Gene Aliases
AT2G39730, RUBISCO ACTIVASE, T5I7.18, rubisco activase
Chromosome Location
Chr.2: 16570746 - 16573692 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129531.3 NP_565913.1 4 - 5 903 70
NM_179989.3 NP_850320.1 4 - 5 903 70
NM_179990.1 NP_850321.1 4 - 5 730 70
GenBank mRNA AF325049.2 - 4 - 5 644 70
AY052703.1 - 4 - 5 633 70
AY056108.1 - 4 - 5 644 70
AY088487.1 - 4 - 5 759 70
BT000710.1 - 4 - 5 641 70

View Full Product Details