Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
XIB
Entrez Gene ID
839246
Gene Name
myosin XI B
Gene Aliases
ARABIDOPSIS THALIANA MYOSIN XI B, AT1G04160, ATXIB, F20D22.7, F20D22_7, MYOSIN XI B, MYOSIN XI-8, XI-8, XI-B, myosin XI B
Chromosome Location
Chr.1: 1086170 - 1096510 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001331465.1 NP_001321769.1 28 - 29 3620 149
NM_100297.3 NP_171912.2 28 - 29 3620 149

View Full Product Details