Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACX6
Entrez Gene ID
837141
Gene Name
acyl-CoA oxidase 6
Gene Aliases
AT1G06310, T2D23.2, T2D23_2, acyl-CoA oxidase 6
Chromosome Location
Chr.1: 1926652 - 1930397 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100512.3 NP_172120.2 3 - 4 847 139

View Full Product Details