Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NDA1
Entrez Gene ID
837229
Gene Name
alternative NAD(P)H dehydrogenase 1
Gene Aliases
ARABIDOPSIS THALIANA INTERNAL NON-PHOSPHORYLATING NAD ( P ) H DEHYDROGENASE, AT1G07180, ATNDI1, F10K1.11, F10K1_11, NADH-DEHYDROGENASE, alternative NAD(P)H dehydrogenase 1, alternative NAD(P)H dehydrogenase 1
Chromosome Location
Chr.1: 2204104 - 2207099 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100592.5 NP_563783.1 5 - 6 1392 82
GenBank mRNA AK118982.1 - 5 - 6 1176 82
BT005564.1 - 5 - 6 1082 82

View Full Product Details