Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRFL2
Entrez Gene ID
837268
Gene Name
TRF-like 2
Gene Aliases
AT1G07540, F22G5.8, F22G5_8, TRF-like 2
Chromosome Location
Chr.1: 2318143 - 2321664 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001331707.1 NP_001318943.1 6 - 7 1351 149

View Full Product Details