Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TOP6B
- Entrez Gene ID
-
821626
- Gene Name
- topoisomerase 6 subunit B
- Gene Aliases
-
AT3G20780, ATTOP6B, BIN3, BRASSINOSTEROID INSENSITIVE 3, ELONGATED HYPOCOTYL 6, HARLEQUIN, HLQ, HYP6, RHL3, ROOT HAIRLESS 3, topoisomerase 6 subunit B, topoisomerase 6 subunit B
- Chromosome Location
-
Chr.3: 7263900 - 7268750 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_112969.3 |
NP_188714.2 |
8 - 9 |
949 |
127 |
GenBank mRNA |
AJ297843.1 |
- |
8 - 9 |
771 |
127 |
View Full Product Details