Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SIG2
Entrez Gene ID
837376
Gene Name
RNApolymerase sigma subunit 2
Gene Aliases
ABC1, AT1G08540, ATSIG1, ATSIG2, PUTATIVE PLASTID RNA POLYMERASE SIGMA-SUBUNIT, RNA POLYMERASE SIGMA SUBUNIT 1, RNApolymerase sigma subunit 2, SIG1, SIGA, SIGB, SIGMA FACTOR 1, SIGMA FACTOR 2, SIGMA FACTOR B, T27G7.22
Chromosome Location
Chr.1: 2703295 - 2706860 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100727.4 NP_172330.1 5 - 6 1394 81
GenBank mRNA AB004293.1 - 5 - 6 1350 81
AB004820.1 - 5 - 6 1315 81
BT030346.1 - 5 - 6 1228 81
Y15362.1 - - 1279 81

View Full Product Details