Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ECT11
Entrez Gene ID
837511
Gene Name
evolutionarily conserved C-terminal region 11
Gene Aliases
AT1G09810, F21M12.20, F21M12_20, evolutionarily conserved C-terminal region 11
Chromosome Location
Chr.1: 3180926 - 3184224 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001331853.1 NP_001320712.1 4 - 5 1290 127
NM_001331854.1 NP_001320713.1 7 - 8 1488 127
NM_100854.5 NP_172452.3 8 - 9 1407 127
GenBank mRNA AK221696.1 - 8 - 9 1372 127
AK226991.1 - 8 - 9 1409 127
BT029759.1 - 4 - 5 1121 127

View Full Product Details