Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SUT4
Entrez Gene ID
837530
Gene Name
sucrose transporter 4
Gene Aliases
AT1G09960, ATSUC4, ATSUT4, F21M12.35, F21M12_35, SUC4, SUCROSE TRANSPORTER 4, sucrose transporter 4
Chromosome Location
Chr.1: 3243986 - 3247216 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100870.3 NP_172467.1 4 - 5 1648 59
GenBank mRNA AJ289166.1 - 4 - 5 1376 59
AY072092.1 - 4 - 5 1421 59
BT004418.1 - 4 - 5 1376 59

View Full Product Details