Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GSTU18
Entrez Gene ID
837575
Gene Name
glutathione S-transferase TAU 18
Gene Aliases
AT1G10360, ATGSTU18, F14N23.24, F14N23_24, GLUTATHIONE S-TRANSFERASE 29, GST29, glutathione S-transferase TAU 18
Chromosome Location
Chr.1: 3395551 - 3396953 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100910.5 NP_172507.1 1 - 2 468 76
GenBank mRNA AF288190.1 - 1 - 2 321 76
AK317183.1 - 1 - 2 343 76
BT000940.1 - 1 - 2 321 76

View Full Product Details