Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WRKY2
Entrez Gene ID
835726
Gene Name
WRKY DNA-binding protein 2
Gene Aliases
ARABIDOPSIS THALIANA WRKY DNA-BINDING PROTEIN 2, AT5G56270, ATWRKY2, MXK23.1, MXK23_1, WRKY DNA-binding protein 2
Chromosome Location
Chr.5: 22779614 - 22783426 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125010.3 NP_200438.1 3 - 4 2048 123
GenBank mRNA AF418308.1 - 2 - 3 1366 123
AY096493.1 - 3 - 4 1960 123
AY123006.1 - 2 - 3 1366 123

View Full Product Details