Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YKT61
Entrez Gene ID
835918
Gene Name
SNARE-like superfamily protein
Gene Aliases
AT5G58060, ATGP1, ATYKT61, K21L19.5, K21L19_5
Chromosome Location
Chr.5: 23498139 - 23500384 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125191.4 NP_200614.1 1 - 2 334 66
GenBank mRNA AF325040.2 - 1 - 2 326 66
AF385703.1 - 1 - 2 326 66
AY133669.1 - 1 - 2 196 66
U64918.1 - 1 - 2 263 66

View Full Product Details