Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LAC16
Entrez Gene ID
836008
Gene Name
laccase 16
Gene Aliases
AT5G58910, K19M22.11, K19M22_11, laccase 16
Chromosome Location
Chr.5: 23789363 - 23792382 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345328.1 NP_001330402.1 5 - 6 1841 97
NM_001345329.1 NP_001330401.1 5 - 6 1568 97
NM_125281.1 NP_200699.1 4 - 5 1439 97

View Full Product Details