Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARF4
Entrez Gene ID
836166
Gene Name
auxin response factor 4
Gene Aliases
AT5G60450, MUF9.7, MUF9_7, auxin response factor 4
Chromosome Location
Chr.5: 24308255 - 24312784 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345418.1 NP_001331788.1 4 - 5 1145 70
NM_125438.4 NP_200853.1 3 - 4 1217 70
GenBank mRNA AF013466.1 - 3 - 4 675 70
AF344313.1 - 3 - 4 620 70
AY080832.1 - 3 - 4 977 70
AY113977.1 - 3 - 4 620 70

View Full Product Details