Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PFK4
Entrez Gene ID
836279
Gene Name
phosphofructokinase 4
Gene Aliases
AT5G61580, K11J9.3, K11J9_3, phosphofructokinase 4
Chromosome Location
Chr.5: 24761041 - 24764168 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037043.1 NP_001032120.1 10 - 11 1268 129
NM_125551.4 NP_200966.2 10 - 11 1318 129
GenBank mRNA AY099694.1 - 10 - 11 1266 129
AY128873.1 - 10 - 11 1209 129

View Full Product Details