Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PFK4
Entrez Gene ID
836279
Gene Name
phosphofructokinase 4
Gene Aliases
AT5G61580, K11J9.3, K11J9_3, phosphofructokinase 4
Chromosome Location
Chr.5: 24761041 - 24764168 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037043.1 NP_001032120.1 11 - 12 1329 123
NM_125551.4 NP_200966.2 11 - 12 1379 123
GenBank mRNA AY099694.1 - 11 - 12 1327 123
AY128873.1 - 11 - 12 1270 123

View Full Product Details