Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSFB2A
Entrez Gene ID
836322
Gene Name
heat shock transcription factor B2A
Gene Aliases
ARABIDOPSIS THALIANA HEAT SHOCK TRANSCRIPTION FACTOR B2A, ARABIDOPSIS THALIANA HEAT SHOCK TRANSCRIPTION FACTOR B2A, AT-HSFB2A, AT5G62020, MTG10.7, MTG10_7, heat shock transcription factor B2A, heat shock transcription factor B2A
Chromosome Location
Chr.5: 24915807 - 24917643 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125595.4 NP_201008.2 1 - 2 646 114
GenBank mRNA AB493808.1 - 1 - 2 241 114
AJ251867.1 - 1 - 2 316 114
AK229690.1 - 1 - 2 281 114
BT008542.1 - 1 - 2 241 114
BT008658.1 - 1 - 2 281 114

View Full Product Details