Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LACS2
Entrez Gene ID
841367
Gene Name
long-chain acyl-CoA synthetase 2
Gene Aliases
AT1G49430, F13F21.14, F13F21_14, LATERAL ROOT DEVELOPMENT 2, LRD2, long-chain acyl-CoA synthetase 2
Chromosome Location
Chr.1: 18290942 - 18295905 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333383.1 NP_001321632.1 14 - 15 1460 101
NM_103833.5 NP_175368.2 14 - 15 1497 101
GenBank mRNA AF503752.1 - 14 - 15 1251 101
AY065424.1 - 14 - 15 1395 101
AY094420.1 - 14 - 15 1332 101
BT001970.1 - 14 - 15 1251 101

View Full Product Details