Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NQR
Entrez Gene ID
841391
Gene Name
ARP protein (REF)
Gene Aliases
AT1G49670, F14J22.10, F14J22_10
Chromosome Location
Chr.1: 18380804 - 18386146 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198253.1 NP_001185182.1 10 - 11 1245 84
NM_103855.5 NP_175390.2 10 - 11 1283 84
GenBank mRNA AY062728.1 - 10 - 11 1239 84
AY072306.1 - 10 - 11 1244 84
AY093342.1 - 10 - 11 1158 84
BT008393.1 - 10 - 11 1158 84

View Full Product Details