Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NQR
Entrez Gene ID
841391
Gene Name
ARP protein (REF)
Gene Aliases
AT1G49670, F14J22.10, F14J22_10
Chromosome Location
Chr.1: 18380804 - 18386146 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198253.1 NP_001185182.1 12 - 13 1493 73
NM_103855.5 NP_175390.2 12 - 13 1462 73
GenBank mRNA AY062728.1 - 12 - 13 1418 73
AY072306.1 - 12 - 13 1423 73
AY093342.1 - 12 - 13 1337 73
BT008393.1 - 12 - 13 1337 73

View Full Product Details