Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLT2
Entrez Gene ID
841542
Gene Name
Integrase-type DNA-binding superfamily protein
Gene Aliases
AT1G51190, F11M15.6, F11M15_6, PLETHORA 2
Chromosome Location
Chr.1: 18977176 - 18980618 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_103997.4 NP_175530.2 2 - 3 364 68
GenBank mRNA AY506550.1 - 1 - 2 210 68
BT020277.1 - 2 - 3 242 68
BT020460.1 - 1 - 2 140 68

View Full Product Details