The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001203340.2 | NP_001190269.1 | 4 - 5 | 515 | 144 |
NM_121023.4 | NP_568219.1 | 4 - 5 | 499 | 144 | |
GenBank mRNA | AF424566.1 | - | 4 - 5 | 419 | 144 |
AF428323.1 | - | 4 - 5 | 437 | 144 |